2-(Chlormethyl)-5-tert-butyl-m-xylol

Lieferant: SIA ENAMINE

EN30001563.1G
ENAMEN30001563.1GEA 0 EUR
ENAMEN30001563.1G
2-(Chlormethyl)-5-tert-butyl-m-xylol
2-(Chlormethyl)-5-tert-butyl-m-xylol
Formel: C₁₃H₁₉Cl
Molecular Weight: 210,75 g/mol
CAS-Nummer: 19387-83-8
EINECS: 243-013-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR