2-Chlor-3'-methoxyacetanilid

Lieferant: SIA ENAMINE

Synonyme: 2-Chloro-N-(3-methoxyphenyl)acetamide

EN30001584.1G
ENAMEN30001584.1GEA 0 EUR
ENAMEN30001584.1G
2-Chlor-3'-methoxyacetanilid
2-Chlor-3'-methoxyacetanilid
Formel: C₉H₁₀ClNO₂
Molecular Weight: 199,6342 g/mol
MDL: MFCD00018900
CAS-Nummer: 17641-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR