2-Brom-3'-(trifluormethyl)acetophenon

Lieferant: SIA ENAMINE

Synonyme: 3-(Trifluoromethyl)phenacyl bromide, 2-Bromo-1-(3-(trifluoromethyl)phenyl)ethanone

EN30001884.1G
ENAMEN30001884.1GEA 0 EUR
ENAMEN30001884.1G
2-Brom-3'-(trifluormethyl)acetophenon
2-Brom-3'-(trifluormethyl)acetophenon
Formel: C₉H₆BrF₃O
Molecular Weight: 267,04 g/mol
MDL: MFCD03094283
CAS-Nummer: 2003-10-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR