ω-Chlor-3,4-difluoracetophenon
Lieferant: SIA ENAMINE
Synonyme:
2-Chloro-3',4'-difluoroacetophenone, 2-Chloro-1-(3,4-difluorophenyl)ethanone
EN30001950.1G
ENAMEN30001950.1GEA
0
EUR
ENAMEN30001950.1G
ω-Chlor-3,4-difluoracetophenon
ω-Chlor-3,4-difluoracetophenon
Formel:
C₈H₅ClF₂O Molecular Weight: 190,5745 g/mol |
MDL:
MFCD03966888 CAS-Nummer: 51336-95-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...