2-Chlor-N-methylacetamid

Lieferant: SIA ENAMINE

EN30001998.1G
ENAMEN30001998.1GEA 0 EUR
ENAMEN30001998.1G
2-Chlor-N-methylacetamid
2-Chlor-N-methylacetamid
Formel: C₃H₆ClNO
MDL: MFCD00018913
CAS-Nummer: 96-30-0
EINECS: 202-497-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR