4,5-Dichlor-3(2H)-pyridazinon

Lieferant: SIA ENAMINE

Synonyme: 4,5-Dichloro-3-hydroxypyridazine, 4,5-dichloro-2,3-dihydropyridazin-3-one

EN30002005.1G
ENAMEN30002005.1GEA 0 EUR
ENAMEN30002005.1G
4,5-Dichlor-3(2H)-pyridazinon
4,5-Dichlor-3(2H)-pyridazinon
Formel: C₄H₂Cl₂N₂O
Molecular Weight: 164,98 g/mol
MDL: MFCD00051504
CAS-Nummer: 932-22-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR