ω,3,4-Trichloracetophenon

Lieferant: SIA ENAMINE
Gefahr

Synonyme: 2,3',4'-Trichloracetophenon, 3,4-Dichlorphenacylchlorid, α,3,4-Trichloracetophenon

EN30002022.1G
ENAMEN30002022.1GEA 0 EUR
ENAMEN30002022.1G
ω,3,4-Trichloracetophenon
ω,3,4-Trichloracetophenon
Formel: C₈H₅Cl₃O
Molecular Weight: 223,49 g/mol
MDL: MFCD03970382
CAS-Nummer: 42981-08-8
UN: 1759
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR