3-Amino-N,N-dimethylbenzolsulfonamid

Lieferant: SIA ENAMINE

EN30002110.1G
ENAMEN30002110.1GEA 0 EUR
ENAMEN30002110.1G
3-Amino-N,N-dimethylbenzolsulfonamid
3-Amino-N,N-dimethylbenzolsulfonamid
Formel: C₈H₁₂N₂O₂S
MDL: MFCD00602451
CAS-Nummer: 6274-18-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR