3-(Piperidin-1-ylsulfonyl)anilin

Lieferant: SIA ENAMINE

EN30002113.1G
ENAMEN30002113.1GEA 0 EUR
ENAMEN30002113.1G
3-(Piperidin-1-ylsulfonyl)anilin
3-(Piperidin-1-ylsulfonyl)anilin
Formel: C₁₁H₁₆N₂O₂S
MDL: MFCD02063328
CAS-Nummer: 22184-99-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR