3-(Morpholinosulfonyl)anilin

Lieferant: SIA ENAMINE

EN30002114.1G
ENAMEN30002114.1GEA 0 EUR
ENAMEN30002114.1G
3-(Morpholinosulfonyl)anilin
3-(Morpholinosulfonyl)anilin
Formel: C₁₀H₁₄N₂O₃S
Molecular Weight: 242,300 g/mol
MDL: MFCD02090909
CAS-Nummer: 22184-97-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR