4-(Morpholinosulfonyl)anilin

Lieferant: SIA ENAMINE

EN30002200.1G
ENAMEN30002200.1GEA 0 EUR
ENAMEN30002200.1G
4-(Morpholinosulfonyl)anilin
4-(Morpholinosulfonyl)anilin
Formel: C₁₀H₁₄N₂O₃S
Molecular Weight: 242,300 g/mol
MDL: MFCD00277213
CAS-Nummer: 21626-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR