2-Amino-4-(4-chlorphenyl)thiazol

Lieferant: SIA ENAMINE

Synonyme: 4-(4-Chlorophenyl)thiazol-2-amine

EN30002225.1G
ENAMEN30002225.1GEA 0 EUR
ENAMEN30002225.1G
2-Amino-4-(4-chlorphenyl)thiazol
2-Amino-4-(4-chlorophenyl)thiazole
Formel: C₉H₇ClN₂S
Molecular Weight: 210.69 g/mol
MDL: MFCD00047058
CAS-Nummer: 2103-99-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR