2-Amino-5-methyl-4-phenylthiazol

Lieferant: SIA ENAMINE

EN30002228.1G
ENAMEN30002228.1GEA 0 EUR
ENAMEN30002228.1G
2-Amino-5-methyl-4-phenylthiazol
2-Amino-5-methyl-4-phenylthiazole
Formel: C₁₀H₁₀N₂S
Molecular Weight: 190,2648 g/mol
MDL: MFCD00617257
CAS-Nummer: 30709-67-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR