2-Amino-4-(4-cyanphenyl)thiazol

Lieferant: SIA ENAMINE

Synonyme: 4-(2-Amino-1,3-thiazol-4-yl)benzonitrile

EN30002257.1G
ENAMEN30002257.1GEA 0 EUR
ENAMEN30002257.1G
2-Amino-4-(4-cyanphenyl)thiazol
2-Amino-4-(4-cyanophenyl)thiazole
Formel: C₁₀H₇N₃S
Molecular Weight: 201,25 g/mol
MDL: MFCD03407828
CAS-Nummer: 436151-85-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR