2-Amino-6-brombenzothiazol

Lieferant: SIA ENAMINE

EN30002269.1G
ENAMEN30002269.1GEA 0 EUR
ENAMEN30002269.1G
2-Amino-6-brombenzothiazol
2-Amino-6-bromobenzothiazole
Formel: C₇H₅BrN₂S
Molecular Weight: 229,1 g/mol
MDL: MFCD00152229
CAS-Nummer: 15864-32-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR