3-Amino-1H-indazol

Lieferant: SIA ENAMINE

EN30002352.1G
ENAMEN30002352.1GEA 0 EUR
ENAMEN30002352.1G
3-Amino-1H-indazol
3-Amino-1H-indazole
Formel: C₇H₇N₃
MDL: MFCD00182045
CAS-Nummer: 874-05-5
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR