4-(Isopropylamino)diphenylamin

Lieferant: SIA ENAMINE

Synonyme: N1-Isopropyl-N4-phenylbenzene-1,4-diamine, N-Isopropyl-N'-phenyl-p-phenylenediamine

EN30002412.1G
ENAMEN30002412.1GEA 0 EUR
ENAMEN30002412.1G
4-(Isopropylamino)diphenylamin
4-(Isopropylamino)diphenylamin
Formel: C₁₅H₁₈N₂
Molecular Weight: 226,32 g/mol
MDL: MFCD00026348
CAS-Nummer: 101-72-4
EINECS: 202-969-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR