3-Chlor-4-(difluormethoxy)anilin Hydrochlorid

Lieferant: SIA ENAMINE

EN30002421.1G
ENAMEN30002421.1GEA 0 EUR
ENAMEN30002421.1G
3-Chlor-4-(difluormethoxy)anilin Hydrochlorid
3-Chlor-4-(difluormethoxy)anilin Hydrochlorid
Formel: C₇H₇Cl₂F₂NO
MDL: MFCD08447168
CAS-Nummer: 115700-24-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR