3-Amino-4-(4-morpholino)benzotrifluorid

Lieferant: SIA ENAMINE

Synonyme: 2-(4-Morpholinyl)-5-(trifluoromethyl)aniline, 2-Morpholino-5-(trifluoromethyl)aniline

EN30002435.1G
ENAMEN30002435.1GEA 0 EUR
ENAMEN30002435.1G
3-Amino-4-(4-morpholino)benzotrifluorid
3-Amino-4-(4-morpholino)benzotrifluorid
Formel: C₁₁H₁₃F₃N₂O
Molecular Weight: 246,23 g/mol
MDL: MFCD00042160
CAS-Nummer: 784-57-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR