2-Amino-5-tert-butyl-1,3,4-thiadiazol

Lieferant: SIA ENAMINE

Synonyme: 5-(tert-Butyl)-1,3,4-thiadiazol-2-amine

EN30002535.1G
ENAMEN30002535.1GEA 0 EUR
ENAMEN30002535.1G
2-Amino-5-tert-butyl-1,3,4-thiadiazol
2-Amino-5-tert-butyl-1,3,4-thiadiazol
Formel: C₆H₁₁N₃S
Molecular Weight: 157,24 g/mol
MDL: MFCD00040403
CAS-Nummer: 39222-73-6
EINECS: 254-364-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR