4-Aminophthalimid
Lieferant: SIA ENAMINE
Synonyme:
4-Amino-1,2-benzoldicarbonsäureimid, 4-Aminophthalsäureimid
EN30002536.1G
ENAMEN30002536.1GEA
0
EUR
ENAMEN30002536.1G
4-Aminophthalimid
4-Aminophthalimid
Formel:
C₈H₆N₂O₂ Molecular Weight: 162,15 g/mol Siedepunkt: 447 °C (1013 hPa) Schmelzpunkt: 294 °C |
MDL:
MFCD00041854 CAS-Nummer: 3676-85-5 EINECS: 222-948-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...