5-Amino-4-cyan-1-phenyl-1H-pyrazol-3-acetonitril

Lieferant: SIA ENAMINE

Synonyme: 5-Amino-3-(cyanomethyl)-1-phenyl-1H-pyrazole-4-carbonitrile

EN30002690.1G
ENAMEN30002690.1GEA 0 EUR
ENAMEN30002690.1G
5-Amino-4-cyan-1-phenyl-1H-pyrazol-3-acetonitril
5-Amino-4-cyan-1-phenyl-1H-pyrazol-3-acetonitril
Formel: C₁₂H₉N₅
Molecular Weight: 223,2334 g/mol
MDL: MFCD00020729
CAS-Nummer: 7152-40-1
EINECS: 230-496-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR