Ethyl-2-amino-4-phenyl-3-thiophencarboxylat

Lieferant: SIA ENAMINE

Synonyme: Ethyl 2-Amino-4-phenylthiophene-3-carboxylate

EN30002862.1G
ENAMEN30002862.1GEA 0 EUR
ENAMEN30002862.1G
Ethyl-2-amino-4-phenyl-3-thiophencarboxylat
Ethyl-2-amino-4-phenyl-3-thiophencarboxylat
Formel: C₁₃H₁₃NO₂S
Molecular Weight: 247,32 g/mol
MDL: MFCD00126391
CAS-Nummer: 4815-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR