Benzothiazol-2-ylmethanol

Lieferant: SIA ENAMINE

Synonyme: 2-(Hydroxymethyl)benzothiazole, 2-Benzothiazolemethanol, 2-Hydroxymethylbenzothiazole

EN30003166.1G
ENAMEN30003166.1GEA 0 EUR
ENAMEN30003166.1G
Benzothiazol-2-ylmethanol
Benzothiazol-2-ylmethanol
Formel: C₈H₇NOS
Molecular Weight: 165,21 g/mol
MDL: MFCD00226293
CAS-Nummer: 37859-42-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR