2-Mercapto-5-nitrobenzimidazol

Lieferant: SIA ENAMINE

Synonyme: 2-Mercapto-5-nitro-1H-benzimidazol, 5-Nitrobenzimidazol-2-thiol

EN30003266.1G
ENAMEN30003266.1GEA 0 EUR
ENAMEN30003266.1G
2-Mercapto-5-nitrobenzimidazol
2-Mercapto-5-nitrobenzimidazole
Formel: C₇H₅N₃O₂S
Molecular Weight: 195,2 g/mol
Schmelzpunkt: 285…287 °C
MDL: MFCD00804727
CAS-Nummer: 6325-91-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR