3-(Trifluormethyl)phenylhydrazin

Lieferant: SIA ENAMINE

Synonyme: 3-Hydrazinobenzotrifluorid, 1-(3-(Trifluormethyl)phenyl)hydrazin

EN30003549.1G
ENAMEN30003549.1GEA 0 EUR
ENAMEN30003549.1G
3-(Trifluormethyl)phenylhydrazin
3-(Trifluormethyl)phenylhydrazin
Formel: C₇H₇F₃N₂
Molecular Weight: 176,14 g/mol
MDL: MFCD00025093
CAS-Nummer: 368-78-5
EINECS: 206-713-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR