2,4-Difluorphenylhydraziniumchlorid

Lieferant: SIA ENAMINE

Synonyme: 1-(2,4-Difluorphenyl)hydrazin Hydrochlorid, 2,4-Difluorphenylhydrazinhydrochlorid, 2,4-Difluorphenylhydrazin Hydrochlorid

EN30003626.1G
ENAMEN30003626.1GEA 0 EUR
ENAMEN30003626.1G
2,4-Difluorphenylhydraziniumchlorid
2,4-Difluorphenylhydraziniumchlorid
Formel: C₆H₇ClF₂N₂
Molecular Weight: 180,58 g/mol
MDL: MFCD00013383
CAS-Nummer: 51523-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR