2-Phenyl-4-chinolincarbonsäurehydrazid
Lieferant: SIA ENAMINE
EN30003627.1G
ENAMEN30003627.1GEA
0
EUR
ENAMEN30003627.1G
2-Phenyl-4-chinolincarbonsäurehydrazid
2-Phenylquinoline-4-carboxylic acid hydrazide
Formel:
C₁₆H₁₃N₃O |
MDL:
MFCD00160604 CAS-Nummer: 4779-54-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...