Pyridine-2-carbohydrazide

Lieferant: SIA ENAMINE

EN30003839.1G
ENAMEN30003839.1GEA 0 EUR
ENAMEN30003839.1G
Pyridine-2-carbohydrazide
Pyridine-2-carbohydrazide
Formel: C₆H₇N₃O
MDL: MFCD00059782
CAS-Nummer: 1452-63-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR