3-(Dimethylamino)benzhydrazid

Lieferant: SIA ENAMINE

EN30003855.1G
ENAMEN30003855.1GEA 0 EUR
ENAMEN30003855.1G
3-(Dimethylamino)benzhydrazid
3-(Dimethylamino)benzhydrazid
Formel: C₉H₁₃N₃O
MDL: MFCD00238588
CAS-Nummer: 205927-64-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR