N-(4-Chlorphenyl)maleimid

Lieferant: SIA ENAMINE

EN30003895.1G
ENAMEN30003895.1GEA 0 EUR
ENAMEN30003895.1G
N-(4-Chlorphenyl)maleimid
N-(4-Chlorphenyl)maleimid
Formel: C₁₀H₆ClNO₂
CAS-Nummer: 1631-29-4
EINECS: 216-632-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR