4-Amino-3-nitrobenzoesäure

Lieferant: SIA ENAMINE
Achtung

Synonyme: 4-Amino-3-nitrobenzoic Acid

EN30003925.1G
ENAMEN30003925.1GEA 0 EUR
ENAMEN30003925.1G
4-Amino-3-nitrobenzoesäure
4-Amino-3-nitrobenzoic acid
Formel: C₇H₆N₂O₄
Molecular Weight: 182,14 g/mol
Schmelzpunkt: 290 °C
MDL: MFCD00017009
CAS-Nummer: 1588-83-6
EINECS: 216-453-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR