N-(2-Bromphenyl)maleimid

Lieferant: SIA ENAMINE

Synonyme: N-(2-Bromphenyl)maleinsäureimid

EN30003994.1G
ENAMEN30003994.1GEA 0 EUR
ENAMEN30003994.1G
N-(2-Bromphenyl)maleimid
N-(2-Bromphenyl)maleimid
Formel: C₁₀H₆BrNO₂
Molecular Weight: 252,07 g/mol
MDL: MFCD00175422
CAS-Nummer: 36817-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR