2-Chlorbenzamidoxim

Lieferant: SIA ENAMINE

EN30003998.1G
ENAMEN30003998.1GEA 0 EUR
ENAMEN30003998.1G
2-Chlorbenzamidoxim
2-Chlorbenzamidoxim
Formel: C₇H₇ClN₂O
MDL: MFCD00859262
CAS-Nummer: 29568-74-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR