2-Methylbenzamidoxim

Lieferant: SIA ENAMINE

EN30003999.1G
ENAMEN30003999.1GEA 0 EUR
ENAMEN30003999.1G
2-Methylbenzamidoxim
2-Methylbenzamidoxim
Formel: C₈H₁₀N₂O
CAS-Nummer: 40312-14-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR