1-Methylimidazol-2-carbonsäure

Lieferant: SIA ENAMINE

EN30004016.1G
ENAMEN30004016.1GEA 0 EUR
ENAMEN30004016.1G
1-Methylimidazol-2-carbonsäure
1-Methylimidazol-2-carbonsäure
Formel: C₅H₆N₂O₂
MDL: MFCD01863421
CAS-Nummer: 20485-43-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR