Propionohydrazid

Lieferant: SIA ENAMINE

Synonyme: Propionhydrazid, Propionsäurehydrazid

EN30004114.1G
ENAMEN30004114.1GEA 0 EUR
ENAMEN30004114.1G
Propionohydrazid
Propionohydrazid
Formel: C₃H₈N₂O
Molecular Weight: 88.11 g/mol
MDL: MFCD01333200
CAS-Nummer: 5818-15-5
EINECS: 227-390-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR