4-Chlor-3-(trifluormethyl)benzolsulfonylchlorid

Lieferant: SIA ENAMINE

Synonyme: 4-Chlor-3-(trifluormethyl)benzol-1-sulfonylchlorid, 4-Chlor-3-(trifluormethyl)benzolsulfonsäurechlorid, 4-Chlor-3-(trifluormethyl)benzolsulfochlorid

EN30004156.1G
ENAMEN30004156.1GEA 0 EUR
ENAMEN30004156.1G
4-Chlor-3-(trifluormethyl)benzolsulfonylchlorid
4-Chlor-3-(trifluormethyl)benzolsulfonylchlorid
Formel: C₇H₃Cl₂F₃O₂S
Molecular Weight: 279,07 g/mol
MDL: MFCD00024882
CAS-Nummer: 32333-53-2
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR