3-Acetylbenzolsulfonylchlorid

Lieferant: SIA ENAMINE

Synonyme: 3-Acetylbenzol-1-sulfonylchlorid, 3-Acetylbenzolsulfochlorid, 3-Acetylbenzolsulfonsäurechlorid

EN30004283.1G
ENAMEN30004283.1GEA 0 EUR
ENAMEN30004283.1G
3-Acetylbenzolsulfonylchlorid
3-Acetylbenzolsulfonylchlorid
Formel: C₈H₇ClO₃S
Molecular Weight: 218.66
MDL: MFCD03424979
CAS-Nummer: 73035-16-2
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR