2,4-Dimethylbenzolsulfonylchlorid

Lieferant: SIA ENAMINE

Synonyme: 2,4-Dimethylbenzol-1-sulfonylchlorid, m-Xylol-4-sulfonsäurechlorid, 2,4-Dimethylbenzolsulfonsäurechlorid, 2,4-Dimethylbenzolsulfochlorid, m-Xylol-4-sulfonylchlorid

EN30004436.1G
ENAMEN30004436.1GEA 0 EUR
ENAMEN30004436.1G
2,4-Dimethylbenzolsulfonylchlorid
2,4-Dimethylbenzolsulfonylchlorid
Formel: C₈H₉ClO₂S
Molecular Weight: 204,68 g/mol
MDL: MFCD00156143
CAS-Nummer: 609-60-9
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR