1-(2-Bromethoxy)-4-methoxybenzol

Lieferant: SIA ENAMINE

EN30004497.1G
ENAMEN30004497.1GEA 0 EUR
ENAMEN30004497.1G
1-(2-Bromethoxy)-4-methoxybenzol
1-(2-Bromoethoxy)-4-methoxybenzene
Formel: C₉H₁₁BrO₂
MDL: MFCD00017893
CAS-Nummer: 22921-76-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR