3,4-Dimethylbenzolsulfonylchlorid

Lieferant: SIA ENAMINE

Synonyme: o-Xylol-4-sulfonsäurechlorid, 3,4-Dimethylbenzol-1-sulfonylchlorid, o-Xylol-4-sulfonylchlorid, 3,4-Dimethylbenzolsulfochlorid, 3,4-Dimethylbenzolsulfonsäurechlorid

EN30004519.1G
ENAMEN30004519.1GEA 0 EUR
ENAMEN30004519.1G
3,4-Dimethylbenzolsulfonylchlorid
3,4-Dimethylbenzolsulfonylchlorid
Formel: C₈H₉ClO₂S
Molecular Weight: 204,68 g/mol
MDL: MFCD00625742
CAS-Nummer: 2905-30-8
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR