3-Sulfamoylbenzoesäure

Lieferant: SIA ENAMINE

EN30004520.1G
ENAMEN30004520.1GEA 0 EUR
ENAMEN30004520.1G
3-Sulfamoylbenzoesäure
3-Sulfamoylbenzoesäure
Formel: C₇H₇NO₄S
MDL: MFCD01122318
CAS-Nummer: 636-76-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR