2-Amino-3,5-dichlor-4-methylpyridin

Lieferant: SIA ENAMINE

Synonyme: 3,5-Dichloro-4-methylpyridin-2-amine

EN30004611.1G
ENAMEN30004611.1GEA 0 EUR
ENAMEN30004611.1G
2-Amino-3,5-dichlor-4-methylpyridin
2-Amino-3,5-dichloro-4-methylpyridine
Formel: C₆H₆Cl₂N₂
Molecular Weight: 177,0300 g/mol
MDL: MFCD00129031
CAS-Nummer: 31430-47-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR