1-(2-Bromethoxy)-4-methylbenzol

Lieferant: SIA ENAMINE

EN30004672.1G
ENAMEN30004672.1GEA 0 EUR
ENAMEN30004672.1G
1-(2-Bromethoxy)-4-methylbenzol
1-(2-Bromoethoxy)-4-methylbenzene
Formel: C₉H₁₁BrO
CAS-Nummer: 18800-34-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR