2-Chlor-4'-methylacetophenon

Lieferant: SIA ENAMINE

EN30004687.1G
ENAMEN30004687.1GEA 0 EUR
ENAMEN30004687.1G
2-Chlor-4'-methylacetophenon
2-Chlor-4'-methylacetophenon
Formel: C₉H₉ClO
CAS-Nummer: 4209-24-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR