1-(4-Bromphenyl)-2,5-dimethylpyrrol
Lieferant: SIA ENAMINE
Synonyme:
1-(4-Bromophenyl)-2,5-dimethyl-1H-pyrrole
EN30004838.1G
ENAMEN30004838.1GEA
0
EUR
ENAMEN30004838.1G
1-(4-Bromphenyl)-2,5-dimethylpyrrol
1-(4-Bromophenyl)-2,5-dimethylpyrrole
Formel:
C₁₂H₁₂BrN |
MDL:
MFCD00277042 CAS-Nummer: 5044-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...