5-Brom-2-iodbenzoesäure

Lieferant: SIA ENAMINE

EN30004839.1G
ENAMEN30004839.1GEA 0 EUR
ENAMEN30004839.1G
5-Brom-2-iodbenzoesäure
5-Brom-2-iodbenzoesäure
Formel: C₇H₄BrIO₂
MDL: MFCD00079717
CAS-Nummer: 21740-00-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR