3-Amino-3-(3-bromphenyl)propionsäure
Lieferant: SIA ENAMINE
Synonyme:
DL-3-Amino-3-(3-Bromophenyl)propionic acid
EN30004944.1G
ENAMEN30004944.1GEA
0
EUR
ENAMEN30004944.1G
3-Amino-3-(3-bromphenyl)propionsäure
3-Amino-3-(3-bromphenyl)propionsäure
Formel:
C₉H₁₀BrNO₂ Molecular Weight: 244,0852 g/mol |
MDL:
MFCD00995843 CAS-Nummer: 117391-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...