4-(Phenoxymethyl)benzoesäure

Lieferant: SIA ENAMINE

EN30005041.1G
ENAMEN30005041.1GEA 0 EUR
ENAMEN30005041.1G
4-(Phenoxymethyl)benzoesäure
4-(Phenoxymethyl)benzoic acid
Formel: C₁₄H₁₂O₃
MDL: MFCD00623268
CAS-Nummer: 31719-76-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR